Assay Design



Gene Transcripts

Gene Symbol
Ttc13
Entrez Gene ID
234875
Gene Name
tetratricopeptide repeat domain 13
Gene Aliases
BC017545
Chromosome Location
Chr.8: 124671327 - 124722013 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145607.3 NP_663582.2 7 - 8 877 68
GenBank mRNA AK144842.1 - 7 - 8 878 68
AK148102.1 - 9 - 10 1104 68
AK160894.1 - 9 - 10 1115 68
AK189721.1 - 2 - 3 228 68
AK194822.1 - 2 - 3 228 68
AK201148.1 - 2 - 3 221 69
BC017545.1 - 1 - 2 91 68
BC072577.1 - 7 - 8 870 68
BC079851.1 - 8 - 9 932 68

View Full Product Details