Assay Design
Gene Transcripts
- Gene Symbol
- Ribc1
- Entrez Gene ID
-
66611
- Gene Name
- RIB43A domain with coiled-coils 1
- Gene Aliases
-
2610028I09Rik, W08639
- Chromosome Location
-
Chr.X: 152004580 - 152018219 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC050755.1 |
- |
1 - 2 |
123 |
66 |
View Full Product Details