Assay Design



Gene Transcripts

Gene Symbol
Pnliprp1
Entrez Gene ID
18946
Gene Name
pancreatic lipase related protein 1
Gene Aliases
Plrp1
Chromosome Location
Chr.19: 58728887 - 58744169 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018874.2 NP_061362.1 5 - 6 622 63
GenBank mRNA AF061274.1 - 5 - 6 623 63
AK028105.1 - 5 - 6 535 63

View Full Product Details