Assay Design



Gene Transcripts

Gene Symbol
Fastkd2
Entrez Gene ID
75619
Gene Name
FAST kinase domains 2
Gene Aliases
2810421I24Rik
Chromosome Location
Chr.1: 63730599 - 63753385 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172422.3 NP_766010.1 10 - 11 2007 110
GenBank mRNA AK013125.1 - 2 - 3 119 110
AK035322.1 - 10 - 11 2007 110
AK084971.1 - 10 - 11 1982 110
AK129251.1 - 8 - 9 3184 110
AK148224.1 - - 1991 110
AK153916.1 - 9 - 10 1950 110
AK166378.1 - 10 - 11 1992 110
AK167714.1 - 9 - 10 2263 110
BC008271.1 - 9 - 10 1948 110
BC057208.1 - 9 - 10 1941 110
BC080813.1 - 9 - 10 1947 110

View Full Product Details