Assay Design
Gene Transcripts
- Gene Symbol
- Ino80d
- Entrez Gene ID
-
227195
- Gene Name
- INO80 complex subunit D
- Gene Aliases
-
7330405I11, A430093A21Rik, BC010584, OTTMUSG00000002594
- Chromosome Location
-
Chr.1: 63047801 - 63114267 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK133321.1 |
- |
- |
3190 |
59 |
View Full Product Details