Assay Design



Gene Transcripts

Gene Symbol
Kctd18
Entrez Gene ID
51960
Gene Name
potassium channel tetramerisation domain containing 18
Gene Aliases
4932411A20Rik, 6530404F10Rik
Chromosome Location
Chr.1: 57955101 - 57970084 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159864.1 NP_001153336.1 5 - 6 791 64
NR_027630.1 5 - 6 823 64
GenBank mRNA BC075692.1 - 5 - 6 823 64

View Full Product Details