Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175370.4 | NP_780579.1 | 3 - 4 | 317 | 66 |
GenBank mRNA | AK016678.1 | - | 3 - 4 | 475 | 66 |
AK077190.1 | - | 3 - 4 | 285 | 66 | |
AK133373.1 | - | - | 316 | 66 | |
BC145039.1 | - | 2 - 3 | 226 | 66 | |
BC145040.1 | - | 2 - 3 | 226 | 66 |