Assay Design
Gene Transcripts
- Gene Symbol
- Acadl
- Entrez Gene ID
-
11363
- Gene Name
- acyl-Coenzyme A dehydrogenase, long-chain
- Gene Aliases
-
AA960361, AU018452, C79855, LCAD
- Chromosome Location
-
Chr.1: 66830839 - 66863309 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC027412.1 |
- |
9 - 10 |
1125 |
83 |
View Full Product Details