Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122952.1 | NP_001116424.1 | 9 - 10 | 1524 | 96 |
NM_001122953.1 | NP_001116425.1 | 8 - 9 | 1634 | 96 | |
NM_010905.3 | NP_035035.1 | 9 - 10 | 1763 | 96 | |
GenBank mRNA | AF326553.1 | - | 9 - 10 | 1462 | 96 |
AF326554.1 | - | 9 - 10 | 1462 | 96 | |
AK052204.1 | - | 9 - 10 | 1585 | 96 | |
AK054310.1 | - | 9 - 10 | 1627 | 96 | |
AK137731.1 | - | 9 - 10 | 1626 | 96 | |
BC122880.1 | - | 8 - 9 | 1354 | 96 | |
D90173.1 | - | 9 - 10 | 1601 | 96 | |
D90174.1 | - | 9 - 10 | 1634 | 96 | |
D90175.1 | - | - | 1478 | 96 | |
U57633.1 | - | 9 - 10 | 1418 | 96 | |
Y07690.1 | - | 9 - 10 | 1494 | 96 | |
Y07691.1 | - | 8 - 9 | 1363 | 96 |