Assay Design



Gene Transcripts

Gene Symbol
Tek
Entrez Gene ID
21687
Gene Name
endothelial-specific receptor tyrosine kinase
Gene Aliases
AA517024, Cd202b, Hyk, STK1, Tie-2, Tie2
Chromosome Location
Chr.4: 94739289 - 94874976 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290549.1 NP_001277478.1 15 - 16 2925 79
NM_001290551.1 NP_001277480.1 14 - 15 2775 79
NM_013690.3 NP_038718.2 15 - 16 2928 79
GenBank mRNA BC050824.1 - 14 - 15 2769 79
D13738.1 - 15 - 16 2933 79
S67051.1 - 16 - 17 2588 79
X67553.1 - 15 - 16 2696 79
X71426.1 - 16 - 17 2659 79

View Full Product Details