Assay Design



Gene Transcripts

Gene Symbol
Tek
Entrez Gene ID
21687
Gene Name
endothelial-specific receptor tyrosine kinase
Gene Aliases
AA517024, Cd202b, Hyk, STK1, Tie-2, Tie2
Chromosome Location
Chr.4: 94739289 - 94874976 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290549.1 NP_001277478.1 20 - 21 3449 66
NM_001290551.1 NP_001277480.1 19 - 20 3299 66
NM_013690.3 NP_038718.2 20 - 21 3452 66
GenBank mRNA BC050824.1 - 19 - 20 3293 66
D13738.1 - 20 - 21 3457 66
S67051.1 - 21 - 22 3112 66
X67553.1 - 20 - 21 3220 66
X71426.1 - 21 - 22 3183 66

View Full Product Details