Assay Design



Gene Transcripts

Gene Symbol
Fchsd1
Entrez Gene ID
319262
Gene Name
FCH and double SH3 domains 1
Gene Aliases
A030002D08Rik
Chromosome Location
Chr.18: 37957434 - 37969814 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175684.4 NP_783615.2 2 - 3 117 57
GenBank mRNA AK154732.1 - 2 - 3 151 57
BC052886.1 - 2 - 3 148 57
BC057367.1 - 2 - 3 117 57

View Full Product Details