Assay Design



Gene Transcripts

Gene Symbol
Nab1
Entrez Gene ID
17936
Gene Name
Ngfi-A binding protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 52455849 - 52500448 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008667.3 NP_032693.2 1 - 2 235 79
GenBank mRNA BC005627.1 - 1 - 2 235 79
BC016886.1 - 1 - 2 158 79

View Full Product Details