Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081081.2 | NP_001074550.1 | 10 - 11 | 1458 | 128 |
NM_001113383.1 | NP_001106854.1 | 10 - 11 | 1458 | 128 | |
GenBank mRNA | AK046297.1 | - | 5 - 6 | 380 | 128 |
AK048072.1 | - | 8 - 9 | 636 | 128 | |
AK173039.1 | - | 10 - 11 | 1458 | 128 | |
BC025534.1 | - | 4 - 5 | 242 | 128 |