Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028447.3 | NP_082723.2 | 8 - 9 | 1297 | 115 |
GenBank mRNA | AK004443.1 | - | 8 - 9 | 1297 | 115 |
AK143537.1 | - | 8 - 9 | 1289 | 115 | |
BC014817.1 | - | 8 - 9 | 1213 | 115 |