Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021529.3 | NP_067504.2 | 2 - 3 | 283 | 120 |
GenBank mRNA | AB041577.1 | - | 2 - 3 | 272 | 120 |
AJ238247.2 | - | 1 - 2 | 110 | 120 | |
AK013403.1 | - | 1 - 2 | 79 | 120 | |
AK015746.1 | - | 1 - 2 | 111 | 120 | |
AK017550.1 | - | 1 - 2 | 66 | 120 | |
AK028063.1 | - | 2 - 3 | 248 | 120 | |
AK028254.1 | - | 1 - 2 | 202 | 120 | |
AK033405.1 | - | 2 - 3 | 281 | 120 | |
AK040589.1 | - | 1 - 2 | 111 | 120 | |
AK087572.1 | - | 1 - 2 | 106 | 120 | |
AK088240.1 | - | 1 - 2 | 192 | 120 | |
AK163192.1 | - | - | 282 | 120 | |
AK168298.1 | - | 2 - 3 | 284 | 120 | |
BC024754.1 | - | 2 - 3 | 246 | 120 | |
BC138295.1 | - | 1 - 2 | 79 | 120 | |
BC138296.1 | - | 1 - 2 | 79 | 120 |