Assay Design



Gene Transcripts

Gene Symbol
Rasef
Entrez Gene ID
242505
Gene Name
RAS and EF hand domain containing
Gene Aliases
BC030882, RAB45
Chromosome Location
Chr.4: 73714577 - 73791401 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017427.1 NP_001017427.1 16 - 17 1909 119
GenBank mRNA AK132691.1 - 16 - 17 1889 119
BC030882.1 - 12 - 13 1302 119
BC040766.1 - 12 - 13 1291 119
BC125539.1 - 15 - 16 1723 119
BC137774.1 - 15 - 16 1723 119
BC144901.1 - 15 - 16 1659 119

View Full Product Details