Assay Design



Gene Transcripts

Gene Symbol
Slc2a5
Entrez Gene ID
56485
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 5
Gene Aliases
AI526984, Glut5, Slc5a
Chromosome Location
Chr.4: 150119344 - 150144168 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019741.3 NP_062715.2 8 - 9 1348 79
GenBank mRNA AF161071.1 - 6 - 7 79
AK029720.1 - 8 - 9 1349 79
BC023500.1 - 6 - 7 724 79

View Full Product Details