Assay Design



Gene Transcripts

Gene Symbol
Casz1
Entrez Gene ID
69743
Gene Name
castor zinc finger 1
Gene Aliases
2410019P08Rik, AV096627, BC040081, Cst, D4Ertd432e
Chromosome Location
Chr.4: 148804367 - 148954893 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159344.1 NP_001152816.1 16 - 17 3855 70

View Full Product Details