Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159344.1 | NP_001152816.1 | 6 - 7 | 1691 | 62 |
NM_027195.2 | NP_081471.2 | 6 - 7 | 1691 | 62 | |
GenBank mRNA | AK138981.1 | - | 1 - 2 | 478 | 62 |
AK173341.1 | - | 1 - 2 | 481 | 62 | |
BC075673.1 | - | 6 - 7 | 1693 | 62 |