Assay Design
Gene Transcripts
- Gene Symbol
- Slc35e2
- Entrez Gene ID
-
320541
- Gene Name
- solute carrier family 35, member E2
- Gene Aliases
-
A530082C11Rik, AI957035
- Chromosome Location
-
Chr.4: 155600592 - 155623340 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK145065.1 |
- |
2 - 3 |
248 |
140 |
View Full Product Details