Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146299.1 | NP_001139771.1 | 4 - 5 | 1176 | 83 |
NM_172966.3 | NP_766554.2 | 3 - 4 | 1080 | 83 | |
GenBank mRNA | AK028153.1 | - | 3 - 4 | 576 | 83 |
AK029168.1 | - | 3 - 4 | 1081 | 83 | |
AK033601.1 | - | 4 - 5 | 1220 | 83 | |
BC137955.1 | - | 4 - 5 | 1129 | 83 | |
BC145060.1 | - | 4 - 5 | 1129 | 83 |