Assay Design



Gene Transcripts

Gene Symbol
Casc1
Entrez Gene ID
320662
Gene Name
cancer susceptibility candidate 1
Gene Aliases
A230084G12Rik, Las1
Chromosome Location
Chr.6: 145174872 - 145210970 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177222.4 NP_796196.3 11 - 12 1433 83
GenBank mRNA AK132912.1 - 11 - 12 1657 83
AY423542.1 - 11 - 12 1364 83
AY485607.1 - 11 - 12 1433 83
AY485608.1 - 11 - 12 1433 83
BC072568.1 - 9 - 10 1275 83
BC120743.1 - 11 - 12 1337 83
BC125423.1 - 11 - 12 1337 83
BC144801.1 - 11 - 12 1337 83

View Full Product Details