Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145967.1 | NP_001139439.1 | 2 - 3 | 275 | 168 |
NM_175029.3 | NP_778194.3 | 2 - 3 | 657 | 168 | |
GenBank mRNA | AK134838.1 | - | 2 - 3 | 424 | 168 |
AK156463.1 | - | 2 - 3 | 427 | 168 | |
AK170747.1 | - | 2 - 3 | 414 | 168 | |
BB851037.1 | - | 2 - 3 | 276 | 168 | |
BC058981.1 | - | 2 - 3 | 657 | 168 |