Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009802.2 | NP_033932.2 | 7 - 8 | 881 | 122 |
GenBank mRNA | AF079834.1 | - | 6 - 7 | 971 | 122 |
AF079835.1 | - | 7 - 8 | 908 | 122 | |
AK132599.1 | - | 7 - 8 | 882 | 122 | |
BC018582.1 | - | 7 - 8 | 858 | 122 | |
BC036270.1 | - | 7 - 8 | 864 | 122 | |
BC046495.1 | - | 6 - 7 | 657 | 122 | |
BC049973.1 | - | 7 - 8 | 879 | 122 | |
BC055437.1 | - | 7 - 8 | 867 | 122 |