Assay Design



Gene Transcripts

Gene Symbol
Pnmal2
Entrez Gene ID
434128
Gene Name
PNMA-like 2
Gene Aliases
EG434128
Chromosome Location
Chr.7: 16944682 - 16948828 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099636.2 NP_001093106.1 1 - 1 4092 81

View Full Product Details