Assay Design



Gene Transcripts

Gene Symbol
Acsl5
Entrez Gene ID
433256
Gene Name
acyl-CoA synthetase long-chain family member 5
Gene Aliases
1700030F05Rik, ACS2, ACS5, Facl5
Chromosome Location
Chr.19: 55251862 - 55296628 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027976.2 NP_082252.1 21 - 22 2891 141
GenBank mRNA AI838021.1 - 1 - 2 141
AK006541.1 - 10 - 11 1368 141
AK034942.1 - - 2891 141
AK150542.1 - 21 - 22 2557 141
AK150663.1 - 21 - 22 2557 141
AK151378.1 - 21 - 22 2346 141
AK161024.1 - 21 - 22 2244 141
AK168019.1 - 9 - 10 892 141
AK207048.1 - 1 - 2 141
AK214922.1 - 1 - 2 149 141
BC031544.1 - 21 - 22 2325 141

View Full Product Details