Assay Design
Gene Transcripts
- Gene Symbol
- Pdzk1
- Entrez Gene ID
-
59020
- Gene Name
- PDZ domain containing 1
- Gene Aliases
-
1700023D20Rik, 2610507N21Rik, 4921513F16Rik, AI267131, AI314638, AL022680, D3Ertd537e, Pdzd1, mPDZK1
- Chromosome Location
-
Chr.3: 96830065 - 96870926 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK014879.1 |
- |
1 - 2 |
346 |
135 |
View Full Product Details