Assay Design



Gene Transcripts

Gene Symbol
Pik3c2g
Entrez Gene ID
18705
Gene Name
phosphatidylinositol 3-kinase, C2 domain containing, gamma polypeptide
Gene Aliases
C80387, PI3K-C2-gamma
Chromosome Location
Chr.6: 139614072 - 139657235 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011084.2 NP_035214.2 9 - 10 1336 96
NM_207683.2 NP_997566.1 - 4082 96
GenBank mRNA AB008791.1 - - 4082 96
AB008792.1 - 9 - 10 1331 96
AK054271.1 - 9 - 10 1212 96
BC150813.1 - - 4074 96

View Full Product Details