Assay Design



Gene Transcripts

Gene Symbol
Pik3c2g
Entrez Gene ID
18705
Gene Name
phosphatidylinositol 3-kinase, C2 domain containing, gamma polypeptide
Gene Aliases
C80387, PI3K-C2-gamma
Chromosome Location
Chr.6: 139614072 - 139657235 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011084.2 NP_035214.2 10 - 11 1533 94
NM_207683.2 NP_997566.1 - 4279 94
GenBank mRNA AB008791.1 - - 4279 94
AB008792.1 - 10 - 11 1528 94
AK054271.1 - 10 - 11 1409 94
BC150813.1 - - 4271 94

View Full Product Details