Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145442.2 | NP_663417.1 | 3 - 4 | 568 | 159 |
GenBank mRNA | AK077125.1 | - | 3 - 4 | 489 | 159 |
AK154318.1 | - | 3 - 4 | 497 | 159 | |
AK171164.1 | - | 3 - 4 | 497 | 159 | |
BC002277.1 | - | 3 - 4 | 554 | 159 |