Assay Design



Gene Transcripts

Gene Symbol
Cyp2j12
Entrez Gene ID
242546
Gene Name
cytochrome P450, family 2, subfamily j, polypeptide 12
Gene Aliases
BB197342, Cyp2j12-ps, OTTMUSG00000007939
Chromosome Location
Chr.4: 96099314 - 96141272 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100182.2 NP_001093652.1 4 - 5 792 145

View Full Product Details