Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199151.1 | NP_001186080.1 | 2 - 3 | 245 | 86 |
NM_001199153.1 | NP_001186082.1 | 2 - 3 | 150 | 86 | |
NM_001199154.1 | NP_001186083.1 | 2 - 3 | 420 | 86 | |
NM_026408.4 | NP_080684.3 | 2 - 3 | 245 | 86 | |
GenBank mRNA | AF348447.1 | - | 2 - 3 | 173 | 86 |
AK016941.1 | - | 2 - 3 | 162 | 86 | |
AK044974.1 | - | 2 - 3 | 443 | 86 | |
AK081942.1 | - | 2 - 3 | 421 | 86 | |
AK082079.1 | - | 2 - 3 | 249 | 86 | |
AK132346.1 | - | 2 - 3 | 348 | 86 | |
AK160111.1 | - | 2 - 3 | 249 | 86 | |
BC098489.1 | - | 2 - 3 | 149 | 86 | |
BC130225.1 | - | 2 - 3 | 135 | 86 | |
BC152544.1 | - | 2 - 3 | 133 | 86 |