Assay Design



Gene Transcripts

Gene Symbol
Sncaip
Entrez Gene ID
67847
Gene Name
synuclein, alpha interacting protein (synphilin)
Gene Aliases
2810407O15Rik, 4933427B05Rik, BB104594, SYPH1
Chromosome Location
Chr.18: 52767811 - 52915935 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199151.1 NP_001186080.1 3 - 4 310 99
NM_001199153.1 NP_001186082.1 3 - 4 215 99
NM_001199154.1 NP_001186083.1 3 - 4 485 99
NM_026408.4 NP_080684.3 3 - 4 310 99
GenBank mRNA AF348447.1 - 3 - 4 238 99
AK013040.1 - 1 - 2 43 99
AK016941.1 - 3 - 4 227 99
AK044974.1 - 3 - 4 508 99
AK081942.1 - 3 - 4 486 99
AK082079.1 - 3 - 4 314 99
AK132346.1 - 3 - 4 413 99
AK160111.1 - 3 - 4 314 99
BC098489.1 - 3 - 4 214 99
BC130225.1 - 3 - 4 200 99
BC152544.1 - 3 - 4 198 99

View Full Product Details