Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053141.3 | NP_444371.3 | 1 - 1 | 1509 | 63 |
GenBank mRNA | AK030612.1 | - | 1 - 1 | 1464 | 63 |
AK036267.1 | - | 1 - 1 | 167 | 63 | |
AK036829.1 | - | 1 - 1 | 1440 | 63 | |
AY013777.1 | - | 1 - 1 | 1288 | 63 | |
BC115720.1 | - | 1 - 1 | 1413 | 63 | |
BC116625.1 | - | 1 - 1 | 1413 | 63 |