Assay Design



Gene Transcripts

Gene Symbol
0610009O20Rik
Entrez Gene ID
66839
Gene Name
RIKEN cDNA 0610009O20 gene
Gene Aliases
2700004E22Rik, Dele, Kiaa0141
Chromosome Location
Chr.18: 38250249 - 38262629 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024179.5 NP_077141.2 11 - 12 1435 78
GenBank mRNA AK002437.1 - 11 - 12 1425 78
AK009400.1 - 4 - 5 447 78
AK141675.1 - - 1398 78
AK171837.1 - 11 - 12 1426 78
AK172900.1 - 11 - 12 1360 78

View Full Product Details