Assay Design



Gene Transcripts

Gene Symbol
Cgn
Entrez Gene ID
70737
Gene Name
cingulin
Gene Aliases
6330408J11Rik
Chromosome Location
Chr.3: 94760069 - 94787724 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037711.2 NP_001032800.2 5 - 6 1281 94
NM_001293727.1 NP_001280656.1 5 - 6 1257 94
GenBank mRNA AK018143.1 - 5 - 6 1195 94
AK173148.1 - 5 - 6 1258 94
BC042459.1 - 5 - 6 1227 94
BC052941.1 - 5 - 6 1203 94
BC064474.1 - 5 - 6 1205 94
BC087546.1 - 4 - 5 1356 94
BC108378.1 - - 1241 94

View Full Product Details