Assay Design



Gene Transcripts

Gene Symbol
Slc25a33
Entrez Gene ID
70556
Gene Name
solute carrier family 25, member 33
Gene Aliases
5730438N18Rik, Pnc1
Chromosome Location
Chr.4: 149744036 - 149774267 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027460.2 NP_081736.2 4 - 5 625 108
GenBank mRNA AK157423.1 - 4 - 5 626 108
AK187296.1 - 3 - 4 108

View Full Product Details