Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025570.2 | NP_079846.1 | 3 - 4 | 320 | 72 |
GenBank mRNA | AB049645.1 | - | 3 - 4 | 293 | 72 |
AK004053.1 | - | 3 - 4 | 305 | 72 | |
AK011341.1 | - | 3 - 4 | 321 | 72 | |
AK015199.1 | - | - | 304 | 72 | |
AK168665.1 | - | 3 - 4 | 294 | 72 | |
BC039985.1 | - | 3 - 4 | 290 | 72 |