Assay Design



Gene Transcripts

Gene Symbol
Hdhd2
Entrez Gene ID
76987
Gene Name
haloacid dehalogenase-like hydrolase domain containing 2
Gene Aliases
0610039H12Rik, 3110052N05Rik
Chromosome Location
Chr.18: 76944115 - 76972171 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039201.1 NP_001034290.1 1 - 2 114 96
GenBank mRNA AK147794.1 - - 96 96

View Full Product Details