Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029885.1 | NP_084161.1 | 3 - 4 | 507 | 173 |
GenBank mRNA | AK036692.1 | - | 3 - 4 | 275 | 173 |
AK087979.1 | - | 3 - 4 | 324 | 173 | |
AK141382.1 | - | 3 - 4 | 481 | 173 | |
AK163939.1 | - | 3 - 4 | 269 | 173 | |
BC150160.1 | - | 3 - 4 | 258 | 173 | |
BC150482.1 | - | 3 - 4 | 258 | 173 |