Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029885.1 | NP_084161.1 | 7 - 8 | 1156 | 84 |
NM_172512.2 | NP_766100.1 | 6 - 7 | 1059 | 84 | |
GenBank mRNA | AK030849.1 | - | 6 - 7 | 828 | 84 |
AK036692.1 | - | 6 - 7 | 1640 | 84 | |
AK141382.1 | - | 7 - 8 | 1130 | 84 |