Assay Design



Gene Transcripts

Gene Symbol
Sos2
Entrez Gene ID
20663
Gene Name
son of sevenless homolog 2 (Drosophila)
Gene Aliases
SOS-2, mSOS-2
Chromosome Location
Chr.12: 69583761 - 69681852 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135559.1 NP_001129031.1 12 - 13 2309 63
GenBank mRNA Z11664.1 - 11 - 12 1953 63

View Full Product Details