Assay Design
Gene Transcripts
- Gene Symbol
- Itga10
- Entrez Gene ID
-
213119
- Gene Name
- integrin, alpha 10
- Gene Aliases
-
A630048L14
- Chromosome Location
-
Chr.3: 96645584 - 96664519 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK037222.1 |
- |
3 - 4 |
198 |
70 |
BC115770.1 |
- |
23 - 24 |
2866 |
70 |
View Full Product Details