Assay Design



Gene Transcripts

Gene Symbol
Gsg1
Entrez Gene ID
14840
Gene Name
germ cell-specific gene 1
Gene Aliases
-
Chromosome Location
Chr.6: 135237329 - 135254336 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010352.2 NP_034482.2 1 - 2 203 69
GenBank mRNA CA465575.1 - - 193 69
D87325.1 - 1 - 2 82 69

View Full Product Details