Assay Design
Gene Transcripts
- Gene Symbol
- Hsbp1l1
- Entrez Gene ID
-
66255
- Gene Name
- heat shock factor binding protein 1-like 1
- Gene Aliases
-
1810005K13Rik
- Chromosome Location
-
Chr.18: 80229571 - 80250103 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC046504.1 |
- |
2 - 3 |
223 |
165 |
View Full Product Details