Assay Design



Gene Transcripts

Gene Symbol
Dnajc11
Entrez Gene ID
230935
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 11
Gene Aliases
E030019A03Rik
Chromosome Location
Chr.4: 151933720 - 151981959 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172704.3 NP_766292.2 11 - 12 1281 80
GenBank mRNA AK053156.1 - 11 - 12 1263 80
AK077544.1 - 2 - 3 215 80
AK156793.1 - 9 - 10 3373 80
AK171781.1 - 11 - 12 1262 80
AK172542.1 - 11 - 12 1262 80
AK186669.1 - 2 - 3 197 80
BC079852.1 - 11 - 12 1251 80
BC085257.1 - 11 - 12 1255 80

View Full Product Details