Assay Design



Gene Transcripts

Gene Symbol
Dnajc11
Entrez Gene ID
230935
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 11
Gene Aliases
E030019A03Rik
Chromosome Location
Chr.4: 151933720 - 151981959 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172704.3 NP_766292.2 13 - 14 1413 80
GenBank mRNA AK053156.1 - 13 - 14 1395 80
AK077544.1 - 4 - 5 347 80
AK171781.1 - 13 - 14 1394 80
AK172542.1 - 13 - 14 1394 80
AK179457.1 - 4 - 5 329 80
AK212087.1 - 1 - 2 61 80
BC027574.1 - 1 - 2 63 80
BC079852.1 - 13 - 14 1383 80
BC085257.1 - 13 - 14 1387 80

View Full Product Details