Assay Design



Gene Transcripts

Gene Symbol
Scin
Entrez Gene ID
20259
Gene Name
scinderin
Gene Aliases
AW545522, adseverin
Chromosome Location
Chr.12: 40059769 - 40134228 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146196.1 NP_001139668.1 6 - 7 1010 68
NM_009132.2 NP_033158.2 6 - 7 1010 68
GenBank mRNA AK133484.1 - 6 - 7 966 68
AK136080.1 - 4 - 5 552 68
AK167739.1 - 6 - 7 965 68
AK171530.1 - 6 - 7 996 68
AK173295.1 - 6 - 7 972 68
BC063328.1 - 6 - 7 965 68
U04354.1 - 6 - 7 965 68
Y13971.1 - 6 - 7 965 68

View Full Product Details