Assay Design



Gene Transcripts

Gene Symbol
Spink13
Entrez Gene ID
100038417
Gene Name
serine peptidase inhibitor, Kazal type 13
Gene Aliases
ENSMUSG00000073551, Gm10534, Spink5l3
Chromosome Location
Chr.18: 62605898 - 62741387 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168423.2 NP_001161895.1 4 - 5 550 118
GenBank mRNA AK139149.1 - 4 - 5 551 118

View Full Product Details